EMAIL THIS PAGE TO A FRIEND
Nucleosides, nucleotides & nucleic acids
Novel acyclic amide-conjugated nucleosides and their analogues.
PMID 19219740
Abstract
An effective one-step synthesis of new amide-conjugated nucleosides and their analogues, in the presence of 4-(4,6-dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride (DMT-MM) as the condensing agent, is presented. Substrate subunits carrying carboxylic group were obtained by acidic hydrolysis of Michael-type adducts of various 5-substituted uracil derivatives to methyl acrylate. Amine substrate was synthesized by reduction of 1-(2'-cyanoethyl)thymine with sodium borohydride in the presence of nickel (II) chloride as catalyst. Other applied amine substrates were 5'-amino-5'-deoxythymidine and 5-aminouracil.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A5525
5′-Amino-5′-deoxythymidine
C10H15N3O4
855286
5-Aminouracil, 98%
C4H5N3O2