EMAIL THIS PAGE TO A FRIEND
Carcinogenesis
BTG3 tumor suppressor gene promoter demethylation, histone modification and cell cycle arrest by genistein in renal cancer.
PMID 19221000
Abstract
BTG3/ANA/APRO4 has been reported to be a tumor suppressor gene in some malignancies. It constitutes important negative regulatory mechanism for Src-mediated signaling, a negative regulator of the cell cycle and inhibits transcription factor E2F1. We report that BTG3 is downregulated in renal cancer and that the mechanism of inactivation is through promoter hypermethylation. Quantitative real-time polymerase chain reaction (PCR) showed that BTG3 was downregulated in cancer tissues and cells. Genistein and 5-aza-2'-deoxycytidine (5Aza-C) induced BTG3 messenger RNA (mRNA) expression in A498, ACHN and HEK-293 renal cell carcinoma (RCC) cell lines. Bisulfite-modified PCR and DNA sequencing results showed complete methylation of BTG3 promoter in tumor samples and cancer cell lines. Genistein and 5Aza-C treatment significantly decreased promoter methylation, reactivating BTG3 expression. Chromatin immunoprecipitation assay revealed that genistein and 5Aza-C increased levels of acetylated histones 3, 4, 2H3K4, 3H3K4 and RNA polymerase II at the BTG3 promoter indicative of active histone modifications. Enzymatic assays showed genistein and 5Aza-C decreased DNA Methyltransferase, methyl-CpG-binding domain 2 activity and increased HAT activity. Cell cycle and 3-(4,5-dimethylthiazole-2-yl)-2,5-biphenyl tetrazolium bromide cell proliferation assays showed that genistein has antiproliferative effect on cancer cell growth through induction of cell cycle arrest. This is the first report to show that BTG3 is epigenetically silenced in RCC and can be reactivated by genistein-induced promoter demethylation and active histone modification. Genistein had similar effects to that of 5Aza-C, which is a potent demethylating agent with high toxicity and instability. Genistein being a natural, non-toxic, dietary isoflavone is effective in retarding the growth of RCC cells, making it a promising candidate for epigenetic therapy in renal carcinoma.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06-599
Anti-acetyl-Histone H3 Antibody, Detect acetyl-Histone H3 with Anti-acetyl-Histone H3 Antibody (Rabbit Polyclonal Antibody), that has been shown to work in WB, ICC, ChIP, ChIP-seq.
06-866
Anti-acetyl-Histone H4 Antibody, Use Anti-acetyl-Histone H4 Antibody (Rabbit Polyclonal Antibody) has been published and validated in ChIP, WB, ICC to detect acetyl-Histone H4 also known as H4 histone family member A, histone 1-H4a.
07-030
Anti-dimethyl-Histone H3 (Lys4) Antibody, Use Anti-dimethyl-Histone H3 (Lys4) Antibody (rabbit polyclonal antibody) is published and validated in ChIP, DB, ICC, IF, WB, PIA, ChIP-seq. to detect dimethyl-Histone H3 (Lys4) also known as H3K4me2, Histone H3 (di methyl K4).
07-441
Anti-dimethyl-Histone H3 (Lys9) Antibody, Use Anti-dimethyl-Histone H3 (Lys9) Antibody (Rabbit Polyclonal Antibody) validated in DB, ICC, PIA, WB to detect dimethyl-Histone H3 (Lys9) also known as H3K9me2, Histone H3 (di methyl K9).
05-623
Anti-RNA polymerase II Antibody, clone CTD4H8, Anti-RNA polymerase II Antibody, clone CTD4H8 is a high quality Mouse Monoclonal Antibody for the detection of RNA polymerase II and has been published in more than 40 citations and validated for use in ChIP & WB.
07-473
Anti-trimethyl-Histone H3 (Lys4) Antibody, Anti-trimethyl-Histone H3 (Lys4) Antibody is a rabbit polyclonal antibody for detection of Histone H3 trimethylated at lysine 4. Also known as Anti-H3K4me3, this highly specific and well published antibody has been validated in ChIP, DB, WB, PIA, ChIP-seq.
HPA018400
Anti-BTG3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution