EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Activity profile in vitro of micafungin against Spanish clinical isolates of common and emerging species of yeasts and molds.
PMID 19223630
Abstract
A collection of 2,278 isolates belonging to 86 different fungal species was tested with micafungin and eight other drugs using the EUCAST procedures. Micafungin was active against species of Candida and Aspergillus (even azole-resistant species) as well as Penicillium spp., Scedosporium apiospermum, and Acremonium spp. It was inactive for species of Basidiomycota and Mucorales and for multiresistant species such as those of Fusarium.