EMAIL THIS PAGE TO A FRIEND
Cell and tissue research
Characterization of primary neurospheres generated from mouse ventral rostral hindbrain.
PMID 19224248
Abstract
Serotonergic (5-HT) neurons of the reticular formation play a key role in the modulation of behavior, and their dysfunction is associated with severe neurological and psychiatric disorders, such as depression and schizophrenia. However, the molecular mechanisms underlying the differentiation of the progenitor cells and the specification of the 5-HT phenotype are not fully understood. A primary neurosphere cell-culture system from mouse ventral rostral hindbrain at embryonic day 12 was therefore established. The generated primary neurospheres comprised progenitor cells and fully differentiated neurons. Bromodeoxyuridine incorporation experiments in combination with immunocytochemistry for neural markers revealed the proliferation capacity of the neural multipotent hindbrain progenitors within neurospheres and their ability to differentiate toward the neuronal lineage and serotonergic phenotype. Gene expression analysis by reverse transcription with the polymerase chain reaction showed that the neurospheres were regionally specified, as reflected by the expression of the transcription factors Gata2 and Pet1. Treatment of dissociated primary neurospheres with exogenous Shh significantly increased the number of 5-HT-immunopositive cells compared with controls, whereas neutralization of endogenous Shh significantly decreased the number of 5-HT neurons. Thus, the primary neurosphere culture system presented here allows the expansion of hindbrain progenitor cells and the experimental control of their differentiation toward the serotonergic phenotype. This culture system is therefore a useful model for in vitro studies dealing with the development of 5-HT neurons.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4501470
Anti-5-HT-1B antibody produced in rabbit, affinity isolated antibody
SAB4501472
Anti-5-HT-1E antibody produced in rabbit, affinity isolated antibody
SAB4501473
Anti-5-HT-1F antibody produced in rabbit, affinity isolated antibody
SAB4501475
Anti-5-HT-2B antibody produced in rabbit, affinity isolated antibody
SAB4501477
Anti-5-HT-2C antibody produced in rabbit, affinity isolated antibody
SAB4501479
Anti-5-HT-3A antibody produced in rabbit, affinity isolated antibody
SAB4501482
Anti-5-HT-5A antibody produced in rabbit, affinity isolated antibody
H6648 Hanks’ Balanced Salt solution, Modified, with sodium bicarbonate, without phenol red, calcium chloride and magnesium sulfate, liquid, sterile-filtered, suitable for cell culture
H8264 Hanks’ Balanced Salt solution, Modified, with sodium bicarbonate, without phenol red, liquid, sterile-filtered, suitable for cell culture
H9269 Hanks’ Balanced Salt solution, With sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
H9394 Hanks’ Balanced Salt solution, Modified, with sodium bicarbonate, without calcium chloride and magnesium sulfate, liquid, sterile-filtered, suitable for cell culture
H4385 Hanks’ Balanced Salt solution, 10 ×, Modified, without calcium, magnesium or sodium bicarbonate
H8262 Hanks’ Balanced Salt solution, With sodium bicarbonate, sterile-filtered
55021C Hanks’ Balanced Salt solution, HBSS Modified, with phenol red, without calcium, without magnesium, liquid
55037C Hanks’ Balanced Salt solution, HBSS Modified, with calcium, with magnesium, without phenol red, liquid, suitable for cell culture
L4544 Laminin from human fibroblasts, cell culture derived, liquid, sterile-filtered
L6274 Laminin from human placenta, liquid, BioReagent, suitable for cell culture