EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Synthesis of glucose derivatives modified at the 4-OH as potential chain-terminators of cellulose biosynthesis; herbicidal activity of simple monosaccharide derivatives.
PMID 19262928
Abstract
A series of D-glucose derivatives that have been modified at C-4 were synthesised from D-galactose as potential chain terminators of cellulose biosynthesis. Two compounds displayed herbicidal activity in pre-emergence tests and in addition a cell expansion assay at higher concentrations revealed symptomology of a third compound that was indicative of inhibition of cellulose biosynthesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
521183
2-Ethynyl-α,α,α-trifluorotoluene, 97%
C9H5F3