EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Synthesis and biological evaluation of N4-(hetero)arylsulfonylquinoxalinones as HIV-1 reverse transcriptase inhibitors.
PMID 19269831
Abstract
A series of novel N(4)-(hetero)arylsulfonylquinoxalinone derivatives were prepared in a straight and efficient way. Of all the synthesized compounds, five compounds exhibited potent anti-HIV-1 replication activities with IC(50) value at the level of 10(-7) mol/L. Preliminary structure-activity relationships were studied in details and that will shed light on the discovery of more potent non-nucleoside reverse-transcriptase inhibitors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
544280
2,5-Dichlorothiophene-3-sulfonyl chloride, 97%
C4HCl3O2S2