EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
In vitro susceptibility testing procedures for fosfomycin tromethamine.
PMID 1929270
Abstract
Fosfomycin tromethamine (previously fosfomycin trometamol) is an orally administered fosfomycin which may be used for single-dose therapy of uncomplicated urinary tract infections. Fosfomycin tromethamine, norfloxacin, and trimethoprim-sulfamethoxazole inhibited greater than 90% of 352 bacterial isolates representing 25 different species; trimethoprim and nalidixic acid had narrower spectrums of activity. Strains of Escherichia, Citrobacter, Enterobacter, and Klebsiella species were much more susceptible when glucose-6-phosphate was added to the test medium, but isolates belonging to other genera were not affected.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P5396
Phosphomycin disodium salt, antibacterial MurA inhibitor
C3H5Na2O4P