EMAIL THIS PAGE TO A FRIEND
The Journal of cell biology
Reversion-induced LIM interaction with Src reveals a novel Src inactivation cycle.
PMID 19307596
Abstract
Aberrant Src activation plays prominent roles in cancer progression. However, how Src is activated in cancer cells is largely unknown. Genetic Src-activating mutations are rare and, therefore, are insufficient to account for Src activation commonly found in human cancers. In this study, we show that reversion-induced LIM (RIL), which is frequently lost in colon and other cancers as a result of epigenetic silencing, suppresses Src activation. Mechanistically, RIL suppresses Src activation through interacting with Src and PTPL1, allowing PTPL1-dependent dephosphorylation of Src at the activation loop. Importantly, the binding of RIL to Src is drastically reduced upon Src inactivation. Our results reveal a novel Src inactivation cycle in which RIL preferentially recognizes active Src and facilitates PTPL1-mediated inactivation of Src. Inactivation of Src, in turn, promotes dissociation of RIL from Src, allowing the initiation of a new Src inactivation cycle. Epigenetic silencing of RIL breaks this Src inactivation cycle and thereby contributes to aberrant Src activation in human cancers.