EMAIL THIS PAGE TO A FRIEND
Journal of proteome research
FLEXIQuant: a novel tool for the absolute quantification of proteins, and the simultaneous identification and quantification of potentially modified peptides.
PMID 19344176
Abstract
Measurements of protein abundance and quantitative assessment of multiple post-translational modifications (PTMs) within a single protein are increasingly used to understand the control of protein activity, particularly in metazoan cells. General methods of wide applicability and precision/accuracy for quantitative estimation of protein post-translational regulation are lacking. Protein mass spectrometry has evolved from a high-throughput qualitative technique to a potentially general quantitative tool, but there are still serious limitations in dynamic range and coverage. To address some of these limitations, we introduce a novel MS-based quantitative strategy, FLEXIQuant, (Full-Length Expressed Stable Isotope-labeled Proteins for Quantification), which can track changes in relative peptide abundances as a function of PTM, and determine absolute quantity of a protein from its lysate. We examined two subunits of the anaphase-promoting complex, CDC27 and APC5, as a test of our ability to monitor quantitatively, the PTM status of several peptides over time. We find evidence of differential regulation at different sites, a phenomenon we believe will be very widespread. FLEXIQuant proved itself to be capable of serving as a general quantitative tool.