EMAIL THIS PAGE TO A FRIEND
Biomedical materials (Bristol, England)
Bioreducible polymers for efficient gene and siRNA delivery.
PMID 19349651
Abstract
Bioreducible disulfide linkage-employing drug conjugate has already been approved for drug delivery application, and also has shown immense potential in gene and siRNA transfection. This paper will focus on the recent developments in bioreducible polymeric systems for gene and siRNA delivery application, and will discuss the advantages and challenges associated with reducible polymeric carriers.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
39380
3-(Dimethylamino)-1-propylamine, purum, ≥98.0% (GC)
C5H14N2