EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Design, synthesis and anticancer activities of hybrids of indole and barbituric acids--identification of highly promising leads.
PMID 19398334
Abstract
By combining the structural features of indole and barbituric acid, new hybrid molecules were designed and synthesized. Evaluations of these molecules over 60 cell line panel of human cancer cells have identified two molecules with significant anticancer activities. Dockings of two active molecules in the active sites of COX-2, thymidylate synthase and ribonucleotide reductase indicate their strong interactions with these enzymes.