EMAIL THIS PAGE TO A FRIEND
The Journal of antimicrobial chemotherapy
Nasal decolonization of Staphylococcus aureus with mupirocin: strengths, weaknesses and future prospects.
PMID 19451132
Abstract
Staphylococcus aureus in the nose is a risk factor for endogenous staphylococcal infection. UK guidelines recommend the use of mupirocin for nasal decolonization in certain groups of patients colonized with methicillin-resistant S. aureus (MRSA). Mupirocin is effective at removing S. aureus from the nose over a few weeks, but relapses are common within several months. There are only a few prospective randomized clinical trials that have been completed with sufficient patients, but those that have been reported suggest that clearance of S. aureus from the nose is beneficial in some patient groups for the reduction in the incidence of nosocomial infections. There is no convincing evidence that mupirocin treatment reduces the incidence of surgical site infection. New antibiotics are needed to decolonize the nose because bacterial resistance to mupirocin is rising, and so it will become less effective. Furthermore, a more bactericidal antibiotic than mupirocin is needed, on the grounds that it might reduce the relapse rate, and so clear the patient of MRSA for a longer period of time than mupirocin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1448901
Mupirocin, United States Pharmacopeia (USP) Reference Standard
C26H44O9