EMAIL THIS PAGE TO A FRIEND
British journal of cancer
Properties of a cell line from human adenocarcinoma of the rectum.
PMID 194617
Abstract
A new, highly differentiated line of cells derived from adenocarcinoma of the rectum (HT55) is described. This line is noteworthy for the following features: 1. The role played in its development by the use of UV-inactivated Sendai virus to attach tumour cell clumps to plastic bottles. 2. Evidence that it produces RNA-containing material of density 1-5--1-16 g/ml. 3. Induction of bone formation in the stroma when grown in athymic mice. 4. Stimulation of primary CBA mouse embryo fibroblasts to form a transient nodule when mixed with them and injected into adult CBA mice. The karyotype and growth-cycle characteristics of the line are described.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
85061105
HT55 Cell Line human