EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Cytotoxic activity and inhibitory effect on nitric oxide production of triterpene saponins from the roots of Physospermum verticillatum (Waldst & Kit) (Apiaceae).
PMID 19467877
Abstract
Three triterpene saponins isolated from the roots of Physospermum verticillatum and identified as saikosaponin a (1), buddlejasaponin IV (2), and songarosaponin D (3) were investigated in vitro for their cytotoxic activity against a panel of seven different cancer cell lines including ACHN, C32, Caco-2, COR-L23, A375, A549, and Huh-7D12 cell lines. The hydrolysis of sugar unit was performed on saikosaponin a (1) to obtain saikosapogenin a (4). All isolated saponins exhibited strong cytotoxic activity against COR-L23 cell line with IC(50) values ranged from 0.4 to 0.6 microM. A similar activity was recorded for saikogenin a (4). None of the tested compounds affected the proliferation of skin fibroblasts 142BR suggesting a selective action against cancer cells. Moreover, buddlejasaponin IV (2) and songarosaponin D (3) exerted significant inhibition of NO production in LPS induced RAW 264.7 macrophages with IC(50) of 4.2 and 10.4 microM, respectively.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
92031919
COR-L23
V1377
Vinblastine sulfate salt, powder, ≥96% (HPLC)
C46H58N4O9 · H2SO4