EMAIL THIS PAGE TO A FRIEND
Toxicological sciences : an official journal of the Society of Toxicology
Identification of chemical compounds that induce HIF-1alpha activity.
PMID 19502547
Abstract
Cellular metabolism depends on the availability of oxygen and the major regulator of oxygen homeostasis is hypoxia-inducible factor 1 (HIF-1), a highly conserved transcription factor that plays an essential role in cellular and systemic homeostatic responses to hypoxia. HIF-1 is a heterodimeric transcription factor composed of hypoxia-inducible HIF-1alpha and constitutively expressed HIF-1beta. Under hypoxic conditions, the two subunits dimerize, allowing translocation of the HIF-1 complex to the nucleus where it binds to hypoxia-response elements (HREs) and activates expression of target genes implicated in angiogenesis, cell growth, and survival. The HIF-1 pathway is essential to normal growth and development, and is involved in the pathophysiology of cancer, inflammation, and ischemia. Thus, there is considerable interest in identifying compounds that modulate the HIF-1 signaling pathway. To assess the ability of environmental chemicals to stimulate the HIF-1 signaling pathway, we screened a National Toxicology Program collection of 1408 compounds using a cell-based beta-lactamase HRE reporter gene assay in a quantitative high-throughput screening (qHTS) format. Twelve active compounds were identified. These compounds were tested in a confirmatory assay for induction of vascular endothelial growth factor, a known hypoxia target gene, and confirmed compounds were further tested for their ability to mimic the effect of a reduced-oxygen environment on hypoxia-regulated promoter activity. Based on this testing strategy, three compounds (o-phenanthroline, iodochlorohydroxyquinoline, cobalt sulfate heptahydrate) were confirmed as hypoxia mimetics, whereas two compounds (7-diethylamino-4-methylcoumarin and 7,12-dimethylbenz(a)anthracence) were found to interact with HIF-1 in a manner different from hypoxia. These results demonstrate the effectiveness of qHTS in combination with secondary assays for identification of HIF-1alpha inducers and for distinguishing among inducers based on their pattern of activated hypoxic target genes. Identification of environmental compounds having HIF-1alpha activation activity in cell-based assays may be useful for prioritizing chemicals for further testing as hypoxia-response inducers in vivo.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
03374
Cobalt atomic spectroscopy standard concentrate 10.00 g Co, 10.00 g/L, for 1L standard solution, analytical standard
C6768
Cobalt(II) sulfate heptahydrate, ReagentPlus®, ≥99%
CoO4S · 7H2O
12933
Cobalt(II) sulfate heptahydrate, purum, ≥95%
CoO4S · 7H2O
HUTR001C MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, Housekeeping Control
HUTR002C
MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, Empty Vector Control
HUTR003C MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, Non-target Control 1
HUTR004C MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, Non-target Control 2
HUTR00012 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, ARNTL
HUTR00018 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, RORA
HUTR00022 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, STAT3
HUTR00023 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, TP53
HUTR00030 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, TRIAP1
HUTR00036 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, MPP5
HUTR00041 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, ARFIP1
HUTR00043 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, DHX40
HUTR00046 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, LATS2
HUTR00047 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, RBL2
HUTR00238 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, ATP5G2
HUTR00247 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, NDST3
HUTR00256 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, WNK2
HUTR00264 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, EDF1
HUTR00276 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, XCR1
HUTR00286 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, RAB40A
HUTR00297 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, TTR
HUTR00307 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, IFI27L1
HUTR00317 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, AMDHD2
HUTR00331 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, SHKBP1
HUTR00338 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, CCDC123
HUTR00345 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, CCT6B
HUTR00484 MISSION® 3′UTR Lenti GoClone, Powered by SwitchGear Genomics, 3′UTR, human, TP73