EMAIL THIS PAGE TO A FRIEND
Bulletin of experimental biology and medicine
Role of cardiac glycosides in regulation of the growth of retinal tissue explants.
PMID 19513372
Abstract
We studied the role of cardiac glycosides in the regulation of the growth of retinal tissue explants from 10-12-day-old chicken embryos in organotypic culture. The studied compounds produced a dose-dependent effect on cell proliferation in retinal tissue explants. Ouabain (10(-13)M), strophanthin K (10(-13)M) and digoxin (10(-11)M) significantly stimulated explant growth. It was hypothesized that the physiological role of endogenous ouabain is associated with regulation of tissue modeling.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
30030
Cymarin, ≥96% (HPLC)
C30H44O9