EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Efficient synthesis of the first betulonic acid-acetylene hybrids and their hepatoprotective and anti-inflammatory activity.
PMID 19524443
Abstract
The Sonogashira reaction can be applied for the preparation of acetylenic derivatives of betulonic acid where the triterpenoid moiety can serve as either the halo- or the acetylenic component. This reaction opened access to the first derivatives of betulonic acid containing either the arylethynyl (C[triple bond]C-Ar(Het) or the ethynyl (C[triple bond]CH) moieties. From the fundamental perspective, this work illustrates the possibility of selective Pd-catalyzed cross-coupling at terminal acetylenes in the presence of a terminal alkene. Hepatoprotective and anti-inflammatory properties of selected acetylenic derivatives of betulonic acid were investigated using the CCl4-induced hepatitis and carrageenan-induced edema models, respectively.