EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
The role of amphiphilicity and negative charge in glycoprotein 41 interactions in the hydrophobic pocket.
PMID 19534533
Abstract
The hydrophobic pocket within the coiled coil domain of HIV-1 gp41 is considered to be a hot-spot suitable for small molecule intervention of fusion, although so far it has yielded only microM inhibitors. Previous peptide studies have identified specific hydrophobic interactions and a Lys-Asp salt bridge as contributing to binding affinity in the pocket. Negative charge appears to be critical for activity of small molecules. We have examined the role of charge and amphiphilic character in the interaction by studying a series of short pocket binding peptides differing in charge, helical content, and in the presence or absence of the Lys-Asp salt bridge, and a series of fatty acid salts with varying charge and hydrocarbon length. Quantitative binding analysis revealed that long-range electrostatic forces and a greasy nonspecific hydrophobic interaction were sufficient for microM potency. The results suggest that an extended interaction site may be necessary for higher potency. We examined a region of the coiled coil immediately C-terminal to the pocket and found that specific salt bridge and hydrogen bond networks may reside in this region. Negatively charged groups extended toward or beyond the C-terminus of the pocket could therefore result in improved low molecular weight fusion inhibitors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
106410
1-Hexadecanesulfonic acid sodium salt, 98%
C16H33NaO3S
O8380
1-Octanesulfonic acid sodium salt, ~98%
C8H17NaO3S
O0133
1-Octanesulfonic acid sodium salt, BioXtra
C8H17NaO3S
293946
Octadecyl sulfate sodium salt, technical grade, 93%
C18H37NaO4S
30633
Sodium 1-decanesulfonate, ≥99.0% (T)
C10H21NaO3S
D3412
Sodium 1-decanesulfonate, ~98%
C10H21NaO3S
30631
Sodium 1-decanesulfonate, for ion pair chromatography, ≥99.0%
C10H21NaO3S
D0181
Sodium 1-decanesulfonate, BioXtra, ~98%
C10H21NaO3S
106437
Sodium 1-dodecanesulfonate, ReagentPlus®, ≥99%
C12H25NaO3S
74886
Sodium 1-octanesulfonate solution, for ion pair chromatography, concentrate, ampule
C8H17NaO3S
87191
Sodium 1-tetradecanesulfonate, for ion pair chromatography, ≥99.0% (T)
C14H29NaO3S
71443
Sodium decyl sulfate, for ion pair chromatography, ≥99.0% (T)
C10H21NaO4S
71445
Sodium decyl sulfate, for surfactant analysis, ≥99.0% (T)
C10H21NaO4S
L9755
Sodium dodecanoate, 99-100%
C12H23NaO2
H5269
Sodium hexanesulfonate, ≥98% (elemental analysis)
C6H13NaO3S
H9026
Sodium hexanesulfonate, BioXtra
C6H13NaO3S
C5038
Sodium octanoate, ≥99% (capillary GC)
C8H15NaO2
O4003
Sodium octyl sulfate, ≥95%
C8H17NaO4S
75073
Sodium octyl sulfate, for ion pair chromatography, ≥99.0% (T)
C8H17NaO4S
293938
Tetradecyl sulfate sodium salt, 95%
C14H29NaO4S