EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Cyanoguanidine-based lactam derivatives as a novel class of orally bioavailable factor Xa inhibitors.
PMID 19541481
Abstract
The N,N'-disubstituted cyanoguanidine is an excellent bioisostere of the thiourea and ketene aminal functional groups. We report the design and synthesis of a novel class of cyanoguanidine-based lactam derivatives as potent and orally active FXa inhibitors. The SAR studies led to the discovery of compound 4 (BMS-269223, K(i)=6.5nM, EC(2xPT)=32muM) as a selective, orally bioavailable FXa inhibitor with an excellent in vitro liability profile, favorable pharmacokinetics and pharmacodynamics in animal models. The X-ray crystal structure of 4 bound in FXa is presented and key ligand-protein interactions are discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
525588
5-Amino-2-methylindole, 97%
C9H10N2
D76609
Dicyandiamide, 99%
C2H4N4