EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Identification of new Hsp90 inhibitors by structure-based virtual screening.
PMID 19560353
Abstract
Structure-based virtual screening identified pyrimidine-2,4,6-trione and 4H-1,2,4-triazole-3-thiol as novel scaffolds of Hsp90 ATPase inhibitors. Their binding modes in the ATP-binding pocket of Hsp90 were analyzed using AutoDoc program combined with molecular dynamics (MD) simulations.