EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Synthesis and evaluation of new endomorphin analogues modified at the Pro(2) residue.
PMID 19560919
Abstract
Six new endomorphin analogues, incorporating constrained amino acids in place of native proline have been synthesized. Residues of (S)-azetidine-2-carboxylic acid (Aze), 3,4-dehydro-(S)-proline (Delta(3)Pro), azetidine-3-carboxylic acid (3Aze) and dehydro-alanine (DeltaAla) have been used to prepare [Delta(3)Pro(2)]EM-2 (1), [Aze(2)]EM-1 (2), [Aze(2)]EM-2 (3), [3Aze(2)]EM-1 (4), [3Aze(2)]EM-2 (5) and [DeltaAla(2)]EM-2 (6). Binding assays and functional bioactivities for mu- and delta-receptors are reported. The highest affinity, bioactivity and selectivity are shown by peptides 2 and 3 containing the Aze residue.