EMAIL THIS PAGE TO A FRIEND
Progress in clinical and biological research
Immunochemical and GC/MS analysis of protein adducts: dosimetry studies with ethylene oxide.
PMID 1956952
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
48838
Ethylene oxide solution, certified reference material, 50 mg/mL in methanol
C2H4O