EMAIL THIS PAGE TO A FRIEND
Journal of natural products
Antiplasmodial isoflavanones from the roots of Sophora mollis.
PMID 19572738
Abstract
Six new prenylated isoflavanones named sophoronols A-F (1-6), together with eight phenolic constituents, were isolated from the roots of Sophora mollis. Their structures and stereochemistry were established by 1D and 2D NMR techniques, especially HMBC and NOESY as well as CD results. Componds 3 and 5 exhibited moderate anitplasmodial activity against the CQS D10 strain of Plasmodium falciparum, with IC(50) values of 12.9 and 12.8 microM, respectively.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S2500
Scopoletin, ≥99%
C10H8O4