EMAIL THIS PAGE TO A FRIEND
Journal of hazardous materials
An overview of landfill leachate treatment via activated carbon adsorption process.
PMID 19577363
Abstract
Water scarcity and pollution rank equal to climate change as the most urgent environmental issue for the 21st century. To date, the percolation landfill leachate into the groundwater tables and aquifer systems which poses a potential risk and potential hazards towards the public health and ecosystems, remains an aesthetic concern and consideration abroad the nations. Arising from the steep enrichment of globalization and metropolitan growth, numerous mitigating approaches and imperative technologies have currently drastically been addressed and confronted. Confirming the assertion, this paper presents a state of art review of leachate treatment technologies, its fundamental background studies, and environmental implications. Moreover, the key advance of activated carbons adsorption, its major challenges together with the future expectation are summarized and discussed. Conclusively, the expanding of activated carbons adsorption represents a potentially viable and powerful tool, leading to the superior improvement of environmental conservation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C9157
Activated charcoal, suitable for cell culture, suitable for plant cell culture
C
C6289
Activated charcoal, plant cell culture tested
C
C2764
Activated charcoal, untreated, granular, ≤5 mm
C
C2889
Activated charcoal, untreated, granular, 8-20 mesh
C
161551
Activated charcoal, powder, -100 particle size (mesh), decolorizing
C
292591
Activated charcoal, 4-14 mesh, granular, Norit® PK 3-5
C
242233
Activated charcoal, DARCO®, 4-12 mesh particle size, granular
C
242241
Activated charcoal, DARCO®, 12-20 mesh, granular
C
242268
Activated charcoal, DARCO®, 20-40 mesh particle size, granular
C
242276
Activated charcoal, DARCO®, −100 mesh particle size, powder
C
329428
Activated charcoal, NORIT® ROW 0.8 SUPRA, pellets
C
C3014
Activated charcoal, untreated, granular, 20-60 mesh
C
C3345
Activated charcoal, untreated powder, 100-400 mesh
C
C4386
Activated charcoal, acid-washed with hydrochloric acid
C
C5510
Activated charcoal, acid-washed with phosphoric and sulfuric acids
C
390127
Activated charcoal, purified by resistive heating of graphite
C
05112
Activated charcoal, for GC
C
05105
Activated charcoal, powder
C
31616
Activated charcoal, puriss. p.a., powder
C
675326
Activated charcoal, DARCO® KB-G
C
675342
Activated charcoal, DARCO® KB-WJ
C
80485
Activated charcoal, for the determination of AOX, 50-150 μm particle size
C
C7606
Activated charcoal, meets USP testing specifications
C
97876
Activated Charcoal Norit®, Norit CA1, from wood, chemically activated, powder
C
37771
Activated Charcoal Norit®, Norit GAC 1240W, from coal, for potable water processing, steam activated, granular
C
22874
Activated Charcoal Norit®, Norit PK 1-3, from peat, steam activated, granular
C
29204
Activated Charcoal Norit®, Norit RB3, for gas purification, steam activated, rod
C
51127
Activated Charcoal Norit®, Norit RB4C, steam activated, rod, for the purification of gas
C
53663
Activated Charcoal Norit®, Norit SX ultra, from peat, corresponds U.S. Food chemicals codex (3rd Ed.), steam activated and acid washed, highly purified, powder
C
93067
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
C