EMAIL THIS PAGE TO A FRIEND
Biomarker insights
A Rapid and Simple Bioassay Method for Herbicide Detection.
PMID 19578512
Abstract
Chlamydomonas reinhardtii, a unicellular green alga, has been used in bioassay detection of a variety of toxic compounds such as pesticides and toxic metals, but mainly using liquid culture systems. In this study, an algal lawn-agar system for semi-quantitative bioassay of herbicidal activities has been developed. Sixteen different herbicides belonging to 11 different categories were applied to paper disks and placed on green alga lawns in Petri dishes. Presence of herbicide activities was indicated by clearing zones around the paper disks on the lawn 2-3 days after application. The different groups of herbicides induced clearing zones of variable size that depended on the amount, mode of action, and chemical properties of the herbicides applied to the paper disks. This simple, paper-disk-algal system may be used to detect the presence of herbicides in water samples and act as a quick and inexpensive semi-quantitative screening for assessing herbicide contamination.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36129
Hexazinone, PESTANAL®, analytical standard
C12H20N4O2