EMAIL THIS PAGE TO A FRIEND
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research
Restoration of regenerative osteoblastogenesis in aged mice: modulation of TNF.
PMID 19580462
Abstract
Skeletal changes accompanying aging are associated with both increased risk of fractures and impaired fracture healing, which, in turn, is due to compromised bone regeneration potential. These changes are associated with increased serum levels of selected proinflammatory cytokines, e.g., tumor necrosis factor alpha (TNF-alpha). We have used a unique model of bone regeneration to demonstrate (1) that aged-related deficits in direct bone formation can be restored to young mice by treatment with TNF blockers and (2) that the cyclin-dependent kinase inhibitor p21 is a candidate for mediation of the osteoinhibitory effects of TNF. It has been hypothesized recently that TNF antagonists may represent novel anabolic agents, and we believe that the data presented here represent a successful test of this hypothesis.