EMAIL THIS PAGE TO A FRIEND
Research in veterinary science
Seroprevalence of and risk factors for Toxoplasma gondii in sheep raised in the Jaboticabal microregion, São Paulo State, Brazil.
PMID 19589550
Abstract
Seroprevalence of and risk factors for toxoplasmosis in sheep from different properties in the Jaboticabal microregion, São Paulo State, Brazil were determined. Antibodies to Toxoplasma gondii were found in sera of 52.0% of 488 sheep tested by indirect fluorescent antibody test (IFAT > or =64). T. gondii seropositivity in sheep was significantly associated with gender of the sheep, pasturing system, contact with cats, and the use of mineral supplements and the type of feed.