EMAIL THIS PAGE TO A FRIEND
Journal of natural products
Cardenolide glycosides of Thevetia peruviana and triterpenoid saponins of Sapindus emarginatus as TRAIL resistance-overcoming compounds.
PMID 19594141
Abstract
A screening study for TRAIL resistance-overcoming activity was carried out, and activity-guided fractionations of Thevetia peruviana and Sapindus emarginatus led to the isolation of four cardenolide glycosides (1-4) and four triterpenoid saponins (5-8), respectively. In particular, cardenolide glycosides (1 and 2) from T. peruviana were shown to have a significant reversal effect on TRAIL resistance in human gastric adenocarcinoma cells, and real-time PCR showed that thevefolin (2) enhanced mRNA expression of death receptor 4 (DR4) and DR5. In addition, 1H and 13C NMR characterizations are shown for thevefolin (2) for the first time.