EMAIL THIS PAGE TO A FRIEND
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research
The orphan nuclear receptor SHP is a positive regulator of osteoblastic bone formation.
PMID 19594294
Abstract
The orphan nuclear receptor small heterodimer partner (SHP; NR0B2) interacts with a diverse array of transcription factors and regulates a variety of cellular events such as cell proliferation, differentiation, and metabolism. However, the role of SHP in bone formation has not yet been elucidated. SHP expression is significantly increased during osteoblast differentiation, and its expression is partially regulated by bone morphogenetic protein 2 (BMP-2), which plays an important role in bone formation. In our study, inhibition of SHP expression significantly repressed BMP-2-induced osteoblast differentiation and ectopic bone formation. In accordance with these in vitro and in vivo results, osteoblast differentiation in SHP(-/-) mice primary osteoblasts was significantly repressed, and the mice showed decreased bone mass resulting from decreased numbers of osteoblasts. Finally, SHP physically interacts and forms a complex with runt-related transcription factor 2 (Runx2) on the osteocalcin gene promoter, and overexpression of SHP increased Runx2 transactivity via competition with histone deacetylase 4 (HDAC4), an enzyme that inhibits DNA binding of Runx2 to its target genes. Taken together, these results indicate that SHP acts as a novel positive regulator of bone formation by augmenting osteoblast differentiation through regulation of the transcriptional activity of Runx2.