EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Investigation of the relative cellular permeability of DNA-binding pyrrole-imidazole polyamides.
PMID 19610652
Abstract
Pyrrole-imidazole (Py-Im) polyamides are a group of chemicals that are able to bind specifically to DNA sequences in vitro and in mammalian cells. Using a cell based reporter assay, we investigated the size and linker affects on the cellular permeability of polyamides. We found that the conventional beta-alanine-3,3'-diamino-N-methyldipropylamine (betaDa) linker strongly limited the cellular permeability. We discovered that a short ethylene diamine (Et) linker displayed high cellular permeability. With the improved Et linker, we found that the cellular permeability of polyamides was size-dependent.