EMAIL THIS PAGE TO A FRIEND
Expert opinion on drug metabolism & toxicology
Ampicillin-sulbactam: an update on the use of parenteral and oral forms in bacterial infections.
PMID 19621991
Abstract
Ampicillin-sulbactam has a wide range of antibacterial activity that includes Gram-positive and Gram-negative aerobic and anaerobic bacteria. However, the drug is not active against Pseudomonas aeruginosa and pathogens producing extended-spectrum beta-lactamases. The combination could be considered particularly active against Acinetobacter baumannii infections due to the intrinsic activity of sulbactam. The drug is indicated as empirical therapy for a broad range of community acquired infections supervened in adults or children and is effective in either parenteral (ampicillin-sulbactam) or oral (as a mutual prodrug sultamicillin) form. In clinical trials, sultamicillin has proved clinically and bacteriologically effective in adults and children against a variety of frequently encountered infections, including mild upper and lower respiratory tract infections, urinary tract infections, diabetic foot and skin and soft tissue infections. Furthermore, adverse effects rarely occur with the diarrhoea to represent the most commonly reported. The parenteral ampicillin-sulbactam is indicated for community infections of mild-to-moderate severity acquired infections such as intra-abdominal or gynecological. Moreover, it seems to represent the alternative of choice for the treatment of A. baumannii infections for carbapenem-resistant strains in the nosocomial setting. Thus, ampicillin-sulbactam remains a valuable agent in the physician's armamentarium in the management of adult and pediatric infections.