EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Semi-rigid tripeptide agonists of melanocortin receptors.
PMID 19635667
Abstract
A series of 30 RCO-HfR-NH(2) derivatives show preference for the mouse MC1R vs MC3-5Rs. trans-4-HOC(6)H(4)CH=CHCO-HfR-NH(2) (13) [EC(50) (nM): MC1R 83, MC3R 20500, MC4R 18130 and MC5R 935; ratio 1:246:217:11] is 11 times more potent than the lead compound LK-394 Ph(CH(2))(3)CO-HfR-NH(2) (2) and only 11 times less potent than the native tridecapeptide alpha-MSH at mMC1R. Differences in conformations of 2 and 13 are discussed.