EMAIL THIS PAGE TO A FRIEND
PLoS neglected tropical diseases
Fatal human rabies due to Duvenhage virus from a bat in Kenya: failure of treatment with coma-induction, ketamine, and antiviral drugs.
PMID 19636367