EMAIL THIS PAGE TO A FRIEND
Journal of natural products
Synthesis of 3-deoxypentacyclic triterpene derivatives as inhibitors of glycogen phosphorylase.
PMID 19642687
Abstract
The 3-deoxy-2-keto derivatives 5 and 7 of oleanolic acid (1) and ursolic acid (2), respectively, served as precursors to the synthesis of 35 3-deoxy derivatives of pentacyclic triterpenes. The synthesized compounds were biologically assayed for their inhibitory activity against rabbit muscle glycogen phosphorylase a (GPa). Among this series of compounds, 2alpha-hydroxyurs-12-en-28-oic acid (18) (IC(50) = 1.2 microM) exhibited the most potent activity. Preliminary structure-activity relationship analysis for the 3-deoxy triterpene derivatives as GP inhibitors is also discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G0549
Corosolic acid from Lagerstroemia speciosa, ≥98% (HPLC)
C30H48O4