EMAIL THIS PAGE TO A FRIEND
Vaccine
Improving administration regimens of CyaA-based vaccines using TRAP assays to detect antigen-specific CD8(+) T cells directly ex vivo.
PMID 19647811
Abstract
Vaccination with recombinant adenylate cyclase of Bordetella pertussis (CyaA) carrying antigen is a promising approach to target antigen-presenting cells. We have used Trogocytosis Analysis Protocol (TRAP) assays to monitor immune responses raised by different vaccination regimens with recombinant CyaA carrying the ovalbumin antigen. We find that the intradermal, intramuscular or subcutaneous routes are all superior to intravenous injections, and actually lead to a sufficiently high frequency of reactive CTL to be detected and characterized directly ex vivo by TRAP assay or other standard assays. Finally, for all routes, we find a clear boosting effect upon re-injection of the vaccine.