EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Synthesis and antimicrobial activity of some new N-acyl substituted phenothiazines.
PMID 19651462
Abstract
A series of 2-substituted N-acylphenothiazines were synthesized by using imides, N-carboxymethyl imides and the structures of these newly synthesized compounds were confirmed by spectral and elemental analyses. All new compounds were tested for their antibacterial and antifungal activities. Some compounds showed promising antibacterial and antifungal activities.