EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Discovery of orally active 3-pyridinyl-tropane as a potent nociceptin receptor agonist for the management of cough.
PMID 19678644
Abstract
A series of 3-pyridinyl-tropane analogues based on previously reported compound 1 have been synthesized and shown to bind to the nociceptin receptor with high affinity. From the SAR study and our lead optimization efforts, compound 10 was found to possess potent oral antitussive activity in the capsaicin-induced guinea pig model. The rationale for compound selection and the biological profile of the optimized lead (10) are disclosed.