EMAIL THIS PAGE TO A FRIEND
Drugs
Letrozole: a review of its use in the treatment of postmenopausal women with hormone-responsive early breast cancer.
PMID 19678717
Abstract
Letrozole (Femara) is a third-generation, nonsteroidal aromatase inhibitor. Adjuvant therapy with letrozole is more effective than tamoxifen in postmenopausal women with hormone-responsive early breast cancer, and extended adjuvant therapy with letrozole after the completion of adjuvant tamoxifen therapy is more effective than placebo in this patient population; letrozole is generally well tolerated. Ongoing trials will help answer outstanding questions regarding the optimal duration of letrozole therapy in early breast cancer and its efficacy compared with other third-generation aromatase inhibitors such as anastrozole. In the meantime, letrozole should be considered a valuable option in the treatment of postmenopausal women with hormone-responsive early breast cancer, both as adjuvant and extended adjuvant therapy.