EMAIL THIS PAGE TO A FRIEND
The American journal of pathology
Neuroendocrine cancer-specific up-regulating mechanism of insulin-like growth factor binding protein-2 in small cell lung cancer.
PMID 19679880
Abstract
Small cell lung cancer (SCLC) exhibits insulin-like growth factor-dependent growth. SCLC is the most aggressive among known in vivo lung cancers, whereas in vitro growth of SCLC is paradoxically slow as compared with that of non-SCLC (NSCLC). In this study, we demonstrate that SCLC cells overexpress insulin-like growth factor binding protein (IGFBP)-2 via NeuroD, a neuroendocrine cell-specific transcription factor. Chromatin immunoprecipitation, electrophoretic mobility shift, and IGFBP-2 promoter assays all revealed that NeuroD binds to the E-box in the 5'-untranslated region of IGFBP-2. A NeuroD transgene in both airway epithelial and NSCLC cells up-regulated the transcription of IGFBP-2 and retarded cell growth. Recombinant IGFBP-2 repressed the growth of both airway epithelial and NSCLC cells in a dose-dependent manner. A NeuroD-specific small interfering RNA repressed IGFBP-2 expression in SCLC, and neutralization of IGFBP-2 and an IGFBP-2-specific small interfering RNA increased SCLC cell growth. Pathological samples of SCLC also expressed IGFBP-2 abundantly, as compared with NSCLC, and showed only rare (8%) IGFBP-2 promoter methylation, whereas the IGFBP-2 promoter was methylated in 71% of adenocarcinomas and 29% of squamous cell carcinomas. These findings suggest that 1) SCLC has an IGFBP-2 overexpression mechanism distinct from NSCLC, 2) secreted IGFBP-2 contributes to the slow growth of SCLC in vitro, and 3) the epigenetic alterations in the IGFBP-2 promoter contribute to the striking differences in IGFBP-2 expression between SCLC and NSCLC in vivo.