EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Biochemical basis of immunological and retroviral responses to DNA-targeted cytosine deamination by activation-induced cytidine deaminase and APOBEC3G.
PMID 19684020
Abstract
Activation-induced cytidine deaminase (AID) and APOBEC3G catalyze deamination of cytosine to uracil on single-stranded DNA, thereby setting in motion a regulated hypermutagenic process essential for human well-being. However, if regulation fails, havoc ensues. AID plays a central role in the synthesis of high affinity antibodies, and APOBEC3G inactivates human immunodeficiency virus-1. This minireview highlights biochemical and structural properties of AID and APOBEC3G, showing how studies using the purified enzymes provide valuable insight into the considerably more complex biology governing antibody generation and human immunodeficiency virus inactivation.