EMAIL THIS PAGE TO A FRIEND
Immunopharmacology and immunotoxicology
Gemfibrozil, stretching arms beyond lipid lowering.
PMID 19694602
Abstract
Gemfibrozil is long known for its ability to reduce the level of triglycerides in the blood circulation and to decrease the risk of hyperlipidemia. However, a number of recent studies reveal that apart from its lipid-lowering effects, gemfibrozil can also regulate many other signaling pathways responsible for inflammation, switching of T-helper cells, cell-to-cell contact, migration, and oxidative stress. In this review, we have made an honest attempt to analyze various biological activities of gemfibrozil and associated mechanisms that may help to consider this drug for different human disorders as primary or adjunct therapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
91823
Gemfibrozil, analytical standard
C15H22O3