EMAIL THIS PAGE TO A FRIEND
Toxicology mechanisms and methods
Determination of Sulfonamide Residues in Food by Micellar Liquid Chromatography.
PMID 19696937
Abstract
This paper presents new methods of determination of sulfonamide residues in food products of animal origin, based on liquid chromatography with a micellar mobile phase. The methods employ a technique of direct injection of the sample and preliminary isolation of the analyte by extraction in the liquid-solid and liquid-liquid system. The methods have been characterized by providing the parameters of the calibration curves, the range of linearity, limit of detection, and precision and accuracy of particular determinations. The recovery of the sulfonamides introduced into the food products studied has also been determined.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46908
Sulfisomidin, VETRANAL, analytical standard
C12H14N4O2S