EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Identification of a cell-active non-peptide sirtuin inhibitor containing N-thioacetyl lysine.
PMID 19700324
Abstract
To identify cell-active sirtuin inhibitors containing N-thioacetyl lysine, we synthesized compound 1, which was designed based on the structure of the reported N-ethoxycarbonylacetyl lysine-based sirtuin inhibitor NCS-12k. Compound 1 selectively inhibited SIRT1 in enzyme assays. Compound 1 also caused a dose-dependent increase in p53 acetylation in human colon cancer HCT116 cells, indicating the inhibition of SIRT1 in these cells.