EMAIL THIS PAGE TO A FRIEND
Brain research bulletin
Oral pre-treatment with epigallocatechin gallate in 6-OHDA lesioned rats produces subtle symptomatic relief but not neuroprotection.
PMID 19712727
Abstract
Much recent work is investigating the role of oxidative stress and inflammatory mechanisms in the aetiology of neurodegeneration in Parkinson's disease. The present study evaluated whether the green tea constituent epigallocatechin gallate (EGCG) which has both anti-oxidant and anti-inflammatory properties, exerts neuroprotection and symptomatic effects when administered orally as a pre-treatment prior to 6-hydroxydopamine (6-OHDA) lesions. Groups of rats were given either 1mg/kg, 2mg/kg EGCG or vehicle solution for 14 days. Sham or 6-OHDA surgery was performed on day 11 of the drug administration protocol. Behavioural analysis was conducted before drugs/vehicle solution, again during the treatment period and then repeated at fortnightly intervals for 2 months post-operatively. Whilst some subtle behavioural improvements in postural abnormalities and ability to cross a narrow beam were observed in lesioned rats after EGCG (vs. vehicle) there was no evidence of neuroprotection on post-mortem quantification of degree of nigral dopaminergic neuronal loss when comparing the lesioned groups given the various treatments.