EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Structure-activity relationship of isoform selective inhibitors of Rac1/1b GTPase nucleotide binding.
PMID 19716293
Abstract
The synthesis of a series of berberine, phenantridine and isoquinoline derivatives was realized to explore their Rho GTPase nucleotide inhibitory activity. The compounds were evaluated in a nucleotide binding competition assay against Rac1, Rac1b, Cdc42 and in a cellular Rac GTPase activation assay. The insertion of 19 AA in the splice variant Rac1b is shown to be sufficient to introduce a conformational difference that allows compounds 4, 21, 22, and 26 to exhibit selective inhibition of Rac 1b over Rac1.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S5890
Sanguinarine chloride hydrate, ≥98% (HPLC)
C20H14ClNO4 · xH2O