EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
The first Hes1 dimer inhibitors from natural products.
PMID 19716294
Abstract
In the present study, we developed a high-throughput screening system for small molecule-inhibitors of the basic helix-loop-helix (bHLH) transcriptional repressor factor Hes1. Successful dimerization of Hes1 immobilized on a microplate and fluorophore (Cy3)-labelled Hes1 was confirmed. Using this system, several natural products were identified as the first Hes1 dimer inhibitors. Of these, two compounds which were isolated from myxomycetes (true slime molds) inhibited Hes1 from N box-dependent suppression of the gene expression in C3H10T1/2 cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S3939
Staurosporine aglycone, ≥95%
C20H13N3O