EMAIL THIS PAGE TO A FRIEND
Toxicology and applied pharmacology
Enlightened protein: Fhit tumor suppressor protein structure and function and its role in the toxicity of protoporphyrin IX-mediated photodynamic reaction.
PMID 19716840
Abstract
The Fhit tumor suppressor protein possesses Ap(3)A (diadenosine triphosphate - ApppA) hydrolytic activity in vitro and its gene is found inactive in many pre-malignant states due to gene inactivation. For several years Fhit has been a widely investigated protein as its cellular function still remains largely unsolved. Fhit was shown to act as a molecular 'switch' of cell death via cascade operating on the influence of ATR-Chk1 pathway but also through the mitochondrial apoptotic pathway. Notably, Fhit was reported by our group to enhance the overall eradication effect of porphyrin-mediated photodynamic treatment (PDT). In this review the up-to-date findings on Fhit protein as a tumor suppressor and its role in PDT are presented.