EMAIL THIS PAGE TO A FRIEND
The Journal of neuroscience : the official journal of the Society for Neuroscience
Loss of dendritic HCN1 subunits enhances cortical excitability and epileptogenesis.
PMID 19726656
Abstract
Hyperpolarization-activated cation nonselective 1 (HCN1) plasticity in entorhinal cortical (EC) and hippocampal pyramidal cell dendrites is a salient feature of temporal lobe epilepsy. However, the significance remains undetermined. We demonstrate that adult HCN1 null mice are more susceptible to kainic acid-induced seizures. After termination of these with an anticonvulsant, the mice also developed spontaneous behavioral seizures at a significantly more rapid rate than their wild-type littermates. This greater seizure susceptibility was accompanied by increased spontaneous activity in HCN1(-/-) EC layer III neurons. Dendritic Ih in these neurons was ablated, too. Consequentially, HCN1(-/-) dendrites were more excitable, despite having significantly more hyperpolarized resting membrane potentials (RMPs). In addition, the integration of EPSPs was enhanced considerably such that, at normal RMP, a 50 Hz train of EPSPs produced action potentials in HCN1(-/-) neurons. As a result of this enhanced pyramidal cell excitability, spontaneous EPSC frequency onto HCN1(-/-) neurons was considerably greater than that onto wild types, causing an imbalance between normal excitatory and inhibitory synaptic activity. These results suggest that dendritic HCN channels are likely to play a critical role in regulating cortical pyramidal cell excitability. Furthermore, these findings suggest that the reduction in dendritic HCN1 subunit expression during epileptogenesis is likely to facilitate the disorder.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Z3777
ZD7288 hydrate, ≥98% (HPLC)
C15H20N4 · HCl · xH2O