EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Function-oriented biosynthesis of beta-lactone proteasome inhibitors in Salinispora tropica.
PMID 19746976
Abstract
The natural proteasome inhibitor salinosporamide A from the marine bacterium Salinispora tropica is a promising drug candidate for the treatment of multiple myeloma and mantle cell lymphoma. Using a comprehensive approach that combined chemical synthesis with metabolic engineering, we generated a series of salinosporamide analogues with altered proteasome binding affinity. One of the engineered compounds is equipotent to salinosporamide A in inhibition of the chymotrypsin-like activity of the proteasome yet exhibits superior activity in the cell-based HCT-116 assay.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
441171
(Bromomethyl)cyclobutane, 97%
C5H9Br