EMAIL THIS PAGE TO A FRIEND
Die Pharmazie
[Synthesis and alpha-adrenolytic activities of 2-(1,4-benzodioxan-2-yl- methyl)-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline derivatives].
PMID 1976262
Abstract
The synthesis of the title compounds starting from 2-Chlormethylbenzdioxan and Tetrahydroisochinolines is presented. Their actions on the platelet aggregation and the inhibition of alpha-adrenoceptors at the isolated rabbit aorta and the vas deferens of the guinea pig were investigated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
291919
6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride, 97%
C11H15NO2 · HCl